Assay Details
Target Gene Details
Entrez Gene ID: | 11144 |
Gene Name: | DNA meiotic recombinase 1 |
Gene Aliases: |
DMC1H, LIM15, dJ199H16.1 |
Location: |
Chr.22:38509154-38570286 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DMC1 | NM_001278208.1 | NP_001265137.1 | ||
| NM_007068.3 | NP_008999.2 | |||
| XM_006724112.2 | XP_006724175.1 | |||
| XM_011529834.1 | XP_011528136.1 | |||
| XM_011529835.2 | XP_011528137.1 | |||
| XM_011529837.1 | XP_011528139.1 | |||
| XM_011529838.1 | XP_011528140.1 | |||
| XM_017028542.1 | XP_016884031.1 | |||
| AK292617.1 | ||||
| AK297664.1 | ||||
| BC125163.1 | ||||
| BC125164.1 | ||||
| CR456486.1 | CAG30372.1 | |||
| D63882.1 | BAA09932.1 | |||
| D64108.1 | BAA10970.1 |
More Information
Set Membership: |
|

Genomic Map