Assay Details
Target Gene Details
Entrez Gene ID: | 5017 |
Gene Name: | ovo like transcriptional repressor 1 |
Gene Aliases: |
HOVO1 |
Location: |
Chr.11:65787034-65797219 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OVOL1 | NM_004561.3 | 1 | 151 | NP_004552.2 |
| AK291711.1 | 1 | 86 | ||
| AL702845.1 | 1 | 120 | ||
| BC059408.1 | 1 | 127 | AAH59408.1 | |
| BX648725.1 | 1 | 31 | CAH56127.1 | |
| CV572438.1 | 1 | 105 | ||
| DA585109.1 | 1 | 164 |
More Information
Set Membership: |
|

Genomic Map