Assay Details
Target Gene Details
Entrez Gene ID: | 169714 |
Gene Name: | quiescin sulfhydryl oxidase 2 |
Gene Aliases: |
QSCN6L1, SOXN |
Location: |
Chr.9:136206333-136245841 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QSOX2 | NM_181701.3 | 12 | 3146 | NP_859052.3 |
| AJ318051.1 | 12 | 3146 | CAC85331.1 | |
| AJ420461.1 | ||||
| AK128077.1 | 12 | 2811 | BAC87262.1 | |
| AK128750.1 | 6 | 2294 | ||
| AL834369.1 | 9 | 2637 | CAD39032.1 | |
| BC047604.2 | 13 | 3028 | AAH47604.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040564 | Chr.9:136124480 - 136243370 on Build GRCh38 | Gain |
|
| nsv825173 | Chr.9:135892081 - 136495763 on Build GRCh38 | Gain |
|
| dgv13001n54 | Chr.9:136106764 - 136233010 on Build GRCh38 | Gain |
|
| nsv951202 | Chr.9:135770355 - 136558248 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map