Assay Details
Target Gene Details
Entrez Gene ID: | 9692 |
Gene Name: | KIAA0391 |
Gene Aliases: |
MRPP3, PRORP |
Location: |
Chr.14:35121855-35274078 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA0391 | XM_017021837.1 | 1 | 61 | XP_016877326.1 |
Target Gene Details
Entrez Gene ID: | 55012 |
Gene Name: | protein phosphatase 2 regulatory subunit B''gamma |
Gene Aliases: |
C14orf10, G4-1, G5pr |
Location: |
Chr.14:35085467-35122543 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R3C | NM_001305155.1 | NP_001292084.1 | ||
| NM_001305156.1 | NP_001292085.1 | |||
| NM_017917.3 | NP_060387.2 | |||
| NR_130972.1 | ||||
| XM_005267782.3 | XP_005267839.1 | |||
| XM_017021388.1 | XP_016876877.1 | |||
| AK000651.1 | BAA91308.1 | |||
| AK225447.1 | ||||
| AK225512.1 | ||||
| AK293717.1 | ||||
| AK310700.1 | ||||
| AK311337.1 | ||||
| AY157304.1 | AAO17045.1 | |||
| AY157305.1 | AAO17046.1 | |||
| AY518535.1 | AAT44532.1 | |||
| AY518536.1 | ||||
| AY518537.1 | AAT44533.1 | |||
| BC006823.1 | AAH06823.1 | |||
| BC010293.2 | AAH10293.1 | |||
| BC012563.1 | AAH12563.1 | |||
| BC063438.1 | AAH63438.1 | |||
| BX248043.1 | CAD62352.1 | |||
| DA617813.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564264 | Chr.14:35013591 - 35263163 on Build GRCh38 | Gain |
|
| nsv564265 | Chr.14:35043129 - 35155625 on Build GRCh38 | Loss |
|
| dgv1868n100 | Chr.14:35041579 - 35287111 on Build GRCh38 | Gain |
|
| esv3584249 | Chr.14:35060670 - 35255038 on Build GRCh38 | Gain |
|
| nsv1042625 | Chr.14:35076888 - 35211152 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map