Assay Details
Target Gene Details
Entrez Gene ID: | 84913 |
Gene Name: | atonal bHLH transcription factor 8 |
Gene Aliases: |
HATH6, bHLHa21 |
Location: |
Chr.2:85753786-85791383 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATOH8 | NM_032827.6 | 1 | 127 | NP_116216.2 |
| XM_006712122.3 | 1 | 127 | XP_006712185.1 | |
| XM_011533139.1 | 1 | 127 | XP_011531441.1 | |
| AK074681.1 | BAC11135.1 | |||
| AK096049.1 | ||||
| BC021207.2 | AAH21207.1 | |||
| DA629171.1 | 1 | 127 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829487 | Chr.2:85753476 - 85755998 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map