Assay Details
Target Gene Details
Entrez Gene ID: | 6764 |
Gene Name: | suppression of tumorigenicity 5 |
Gene Aliases: |
DENND2B, HTS1, p126 |
Location: |
Chr.11:8693352-8910951 on Build GRCh38 |
Assay Gene Location: | Within Exon 37 |
More Information
Set Membership: |
|

Genomic Map