Assay Details
Target Gene Details
Entrez Gene ID: | 57477 |
Gene Name: | shroom family member 4 |
Gene Aliases: |
SHAP |
Location: |
Chr.X:50575534-50814167 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHROOM4 | NM_020717.3 | 9 | 4786 | NP_065768.2 |
| NR_027121.1 | 9 | 4786 | ||
| XM_017029682.1 | 10 | 5006 | XP_016885171.1 | |
| XM_017029683.1 | 9 | 4871 | XP_016885172.1 | |
| XM_017029684.1 | 9 | 5023 | XP_016885173.1 | |
| XM_017029686.1 | 8 | 4888 | XP_016885175.1 | |
| AB033028.1 | 9 | 4801 | ||
| BC151240.1 | 9 | 4812 | ||
| BI018520.1 | 1 | 294 |
More Information
Set Membership: |
|

Genomic Map