Assay Details
Target Gene Details
Entrez Gene ID: | 23491 |
Gene Name: | carboxylesterase 3 |
Gene Aliases: |
ES31 |
Location: |
Chr.16:66961229-66975152 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CES3 | NM_001185176.1 | NP_001172105.1 | ||
| NM_001185177.1 | NP_001172106.1 | |||
| NM_024922.5 | NP_079198.2 | |||
| XM_011522975.2 | XP_011521277.1 | |||
| AK000105.1 | ||||
| AK025158.1 | ||||
| AK025389.1 | BAB15123.1 | |||
| AK097538.1 | BAC05093.1 | |||
| AY358609.1 | AAQ88972.1 | |||
| BC053670.1 | AAH53670.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827706 | Chr.16:66894354 - 67009614 on Build GRCh38 | Loss |
|
| dgv526e214 | Chr.16:66940449 - 67031371 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map