Assay Details
Target Gene Details
Entrez Gene ID: | 92259 |
Gene Name: | mitochondrial ribosomal protein S36 |
Gene Aliases: |
DC47, MRP-S36 |
Location: |
Chr.5:69217746-69230158 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS36 | NM_033281.5 | NP_150597.1 | ||
| AA780181.1 | ||||
| AL529370.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5694n100 | Chr.5:69081291 - 69374274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map