Assay Details
Target Gene Details
Entrez Gene ID: | 10550 |
Gene Name: | ADP ribosylation factor like GTPase 6 interacting protein 5 |
Gene Aliases: |
DERP11, GTRAP3-18, HSPC127, JWA, PRAF3, Yip6b, addicsin, hp22, jmx |
Location: |
Chr.3:69084939-69106088 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL6IP5 | NM_006407.3 | 3 | 783 | NP_006398.1 |
| AB052638.1 | 3 | 681 | BAD83603.1 | |
| AB097051.1 | 3 | 764 | BAC77404.1 | |
| AF070523.1 | 3 | 789 | AAC64360.1 | |
| AF115965.1 | 3 | 774 | AAP97237.1 | |
| AF125530.1 | 3 | 775 | AAF17224.1 | |
| AF161476.1 | 3 | 764 | AAF29091.1 | |
| AK074612.1 | 3 | 735 | ||
| AK223566.1 | 3 | 687 | BAD97286.1 | |
| AK301637.1 | 4 | 697 | ||
| AK303148.1 | 3 | 669 | ||
| BC005143.2 | 3 | 706 | AAH05143.1 | |
| BC020797.1 | 3 | 700 | AAH20797.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3596491 | Chr.3:69103966 - 69119758 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map