Assay Details
Target Gene Details
Entrez Gene ID: | 1676 |
Gene Name: | DNA fragmentation factor subunit alpha |
Gene Aliases: |
DFF-45, DFF1, ICAD |
Location: |
Chr.1:10460531-10472556 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DFFA | NM_004401.2 | NP_004392.1 | ||
| NM_213566.1 | 5 | 1349 | NP_998731.1 | |
| AF087573.1 | 5 | 1288 | AAD32953.1 | |
| AK222546.1 | BAD96266.1 | |||
| AK313317.1 | ||||
| BC000037.2 | AAH00037.1 | |||
| BC007112.1 | AAH07112.1 | |||
| BC007721.2 | AAH07721.1 | |||
| BT006980.1 | AAP35626.1 | |||
| U91985.1 | AAC51249.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2764192 | Chr.1:10323676 - 10534129 on Build GRCh38 | Gain |
|
| esv2758920 | Chr.1:10178012 - 10522497 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map