Assay Details
Target Gene Details
Entrez Gene ID: | 222235 |
Gene Name: | F-box and leucine rich repeat protein 13 |
Gene Aliases: |
DRC6, Fbl13 |
Location: |
Chr.7:102699226-103074841 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Target Gene Details
Entrez Gene ID: | 10234 |
Gene Name: | leucine rich repeat containing 17 |
Gene Aliases: |
P37NB |
Location: |
Chr.7:102912897-102945111 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC17 | NM_001031692.2 | NP_001026862.1 | ||
| NM_005824.2 | NP_005815.2 | |||
| XM_005250108.1 | XP_005250165.1 | |||
| AK225701.1 | ||||
| AK226021.1 | ||||
| AK290305.1 | ||||
| AY358889.1 | AAQ89248.1 | |||
| BC027903.1 | AAH27903.1 | |||
| DA749917.1 | ||||
| U32907.1 | AAB41565.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464667 | Chr.7:102845807 - 102983745 on Build GRCh38 | Loss |
|
| dgv6538n100 | Chr.7:102171110 - 103063688 on Build GRCh38 | Gain |
|
| esv2752138 | Chr.7:102717873 - 103146688 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map