Assay Details
Target Gene Details
Entrez Gene ID: | 120776 |
Gene Name: | olfactory receptor family 2 subfamily D member 2 |
Gene Aliases: |
OR11-610, OR2D1, hg27 |
Location: |
Chr.11:6891574-6892500 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR2D2 | NM_003700.1 | NP_003691.1 | ||
| BC140729.1 | 1 | 992 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035138 | Chr.11:6607812 - 6980555 on Build GRCh38 | Gain |
|
| nsv832061 | Chr.11:6847937 - 7049841 on Build GRCh38 | Loss |
|
| nsv1046129 | Chr.11:6689483 - 6941934 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map