Assay Details
Target Gene Details
Entrez Gene ID: | 2299 |
Gene Name: | forkhead box I1 |
Gene Aliases: |
FKH10, FKHL10, FREAC-6, FREAC6, HFH-3, HFH3 |
Location: |
Chr.5:170105897-170109725 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXI1 | NM_012188.4 | 1 | 398 | NP_036320.2 |
| NM_144769.2 | 1 | 398 | NP_658982.1 | |
| AK292176.1 | 1 | 414 | ||
| AY707089.1 | 1 | 272 | AAU12169.1 | |
| BC029778.1 | 1 | 398 | ||
| L13203.1 | 1 | 350 | AAB50574.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830547 | Chr.5:170083822 - 170257632 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map