Assay Details
Target Gene Details
Entrez Gene ID: | 79696 |
Gene Name: | zinc finger C2HC-type containing 1C |
Gene Aliases: |
C14orf140, FAM164C |
Location: |
Chr.14:75069577-75079987 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZC2HC1C | NM_001042430.2 | NP_001035895.1 | ||
| NM_024643.3 | 2 | 1165 | NP_078919.2 | |
| NR_110315.1 | ||||
| XM_005268062.3 | 2 | 1241 | XP_005268119.1 | |
| XM_005268063.4 | XP_005268120.1 | |||
| XM_006720255.3 | 2 | 1101 | XP_006720318.1 | |
| XM_011537155.2 | 2 | 1203 | XP_011535457.1 | |
| XM_011537156.2 | 2 | 1050 | XP_011535458.1 | |
| XM_011537157.2 | 2 | 1101 | XP_011535459.1 | |
| XM_017021643.1 | 2 | 1101 | XP_016877132.1 | |
| XM_017021644.1 | XP_016877133.1 | |||
| XM_017021645.1 | XP_016877134.1 | |||
| AK026746.1 | 2 | 1100 | BAB15541.1 | |
| AK223359.1 | BAD97079.1 | |||
| AK303882.1 | 2 | 1103 | ||
| BC004259.1 | AAH04259.1 | |||
| BX110341.1 |
More Information
Set Membership: |
|

Genomic Map