Assay Details
Target Gene Details
Entrez Gene ID: | 150709 |
Gene Name: | ankyrin and armadillo repeat containing |
Gene Aliases: |
- |
Location: |
Chr.2:189674292-189763059 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKAR | XM_011510676.2 | XP_011508978.1 | ||
| XM_011510677.2 | XP_011508979.1 | |||
| XM_011510681.1 | XP_011508983.1 | |||
| XM_011510682.2 | XP_011508984.1 | |||
| XM_017003413.1 | XP_016858902.1 | |||
| BC044907.1 | AAH44907.1 |
Target Gene Details
Entrez Gene ID: | 64172 |
Gene Name: | O-sialoglycoprotein endopeptidase-like 1 |
Gene Aliases: |
Qri7 |
Location: |
Chr.2:189746660-189763198 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OSGEPL1 | NM_022353.2 | NP_071748.2 | ||
| XM_005246766.3 | XP_005246823.1 | |||
| XM_006712685.1 | XP_006712748.1 | |||
| XM_006712686.1 | XP_006712749.1 | |||
| XM_011511631.1 | XP_011509933.1 | |||
| XM_017004676.1 | XP_016860165.1 | |||
| XM_017004677.1 | XP_016860166.1 | |||
| XM_017004678.1 | XP_016860167.1 | |||
| XM_017004679.1 | XP_016860168.1 | |||
| XM_017004680.1 | XP_016860169.1 | |||
| XM_017004681.1 | XP_016860170.1 | |||
| XM_017004682.1 | XP_016860171.1 | |||
| XM_017004683.1 | XP_016860172.1 | |||
| XM_017004684.1 | XP_016860173.1 | |||
| XM_017004685.1 | 7 | 1409 | XP_016860174.1 | |
| AJ295148.1 | CAC14666.1 | |||
| AK055441.1 | BAB70923.1 | |||
| AK314565.1 | ||||
| BC011904.2 | AAH11904.1 |
More Information
Set Membership: |
|

Genomic Map