Assay Details
Target Gene Details
Entrez Gene ID: | 83743 |
Gene Name: | glutamate rich WD repeat containing 1 |
Gene Aliases: |
CDW4, GRWD, RRB1, WDR28 |
Location: |
Chr.19:48445773-48453907 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRWD1 | NM_031485.3 | 7 | 1477 | NP_113673.3 |
| AF337808.1 | 7 | 1256 | AAK17998.1 | |
| AK074676.1 | 7 | 1445 | BAC11130.1 | |
| AK131055.1 | 6 | 683 | ||
| AK300225.1 | 7 | 930 | ||
| AK315625.1 | 7 | 1269 | ||
| AL713729.1 | 7 | 1252 | CAD28519.1 | |
| BC002440.2 | 7 | 1252 | AAH02440.1 | |
| EF011614.1 | 7 | 1250 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv828572 | Chr.19:48399035 - 48497609 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map