Assay Details
Target Gene Details
Entrez Gene ID: | 57605 |
Gene Name: | phosphatidylinositol transfer protein membrane associated 2 |
Gene Aliases: |
NIR-3, NIR3, RDGB2, RDGBA2 |
Location: |
Chr.12:122983480-123110489 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PITPNM2 | NM_001300801.1 | 25 | 5584 | NP_001287730.1 |
| NM_020845.2 | 25 | 5541 | NP_065896.1 | |
| XM_011538590.2 | 26 | 5863 | XP_011536892.1 | |
| AB040890.2 | 26 | 5605 | ||
| AK026156.1 | 1 | 534 | ||
| AL133612.1 | 16 | 3988 | CAB63741.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv914 | Chr.12:122977362 - 123008698 on Build GRCh38 | Insertion |
|
| dgv22n68 | Chr.12:122911380 - 123104743 on Build GRCh38 | Loss |
|
| nsv560493 | Chr.12:122982778 - 122995076 on Build GRCh38 | Loss |
|
| nsv560492 | Chr.12:122943026 - 123090291 on Build GRCh38 | Loss |
|
| nsv517200 | Chr.12:122827504 - 123169951 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map