Assay Details
Target Gene Details
Entrez Gene ID: | 140461 |
Gene Name: | ankyrin repeat and SOCS box containing 8 |
Gene Aliases: |
PP14212 |
Location: |
Chr.12:48147788-48157595 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB8 | NM_001319296.1 | 5 | 1266 | NP_001306225.1 |
| NM_001319297.1 | 4 | 1126 | NP_001306226.1 | |
| NM_001319298.1 | 5 | 1231 | NP_001306227.1 | |
| NM_001319299.1 | 5 | 1227 | NP_001306228.1 | |
| NM_001319300.1 | 5 | 1184 | NP_001306229.1 | |
| NM_001319301.1 | 5 | 1180 | NP_001306230.1 | |
| NM_024095.4 | 4 | 1130 | NP_077000.1 | |
| AF464877.1 | 4 | 1022 | AAQ04830.1 | |
| AK024908.1 | 4 | 1031 | BAB15033.1 | |
| AK125596.1 | 1 | 1877 | BAC86214.1 | |
| AK289957.1 | 4 | 1097 | ||
| AK297013.1 | 5 | 936 | ||
| BC001321.1 | 4 | 1037 | AAH01321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759898 | Chr.12:48035919 - 48431716 on Build GRCh38 | Gain+Loss |
|
| esv3629429 | Chr.12:48142951 - 48278499 on Build GRCh38 | Gain |
|
| nsv508673 | Chr.12:48096003 - 48173355 on Build GRCh38 | Deletion |
|
| nsv8971 | Chr.12:48142743 - 48364718 on Build GRCh38 | Gain+Loss |
|
| esv34439 | Chr.12:48147974 - 48385779 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map