Assay Details
Target Gene Details
Entrez Gene ID: | 27147 |
Gene Name: | DENN domain containing 2A |
Gene Aliases: |
FAM31D, KIAA1277 |
Location: |
Chr.7:140518418-140642209 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND2A | NM_001318052.1 | NP_001304981.1 | ||
| NM_015689.4 | NP_056504.3 | |||
| NR_134477.1 | ||||
| XM_005249977.3 | XP_005250034.1 | |||
| XM_011516052.2 | XP_011514354.1 | |||
| XM_011516053.2 | XP_011514355.1 | |||
| XM_011516054.2 | XP_011514356.1 | |||
| XM_017011989.1 | XP_016867478.1 | |||
| AB033103.2 | ||||
| AK290056.1 | ||||
| AK297959.1 | ||||
| BC049193.1 | AAH49193.1 | |||
| BC132875.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761112 | Chr.7:140491738 - 140534771 on Build GRCh38 | Gain+Loss |
|
| dgv6642n100 | Chr.7:140497279 - 140535877 on Build GRCh38 | Gain |
|
| nsv608501 | Chr.7:140495663 - 140533785 on Build GRCh38 | Gain |
|
| nsv1018535 | Chr.7:140498740 - 140524503 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map