Assay Details
Target Gene Details
Entrez Gene ID: | 7014 |
Gene Name: | telomeric repeat binding factor 2 |
Gene Aliases: |
TRBF2, TRF2 |
Location: |
Chr.16:69355561-69386004 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TERF2 | NM_005652.4 | 7 | 1236 | NP_005643.2 |
| XM_005256121.3 | 8 | 1261 | XP_005256178.1 | |
| XM_005256122.3 | 8 | 1261 | XP_005256179.1 | |
| XM_005256123.3 | 7 | 1252 | XP_005256180.1 | |
| XM_005256124.3 | 5 | 1018 | XP_005256181.1 | |
| AF002999.1 | 7 | 1217 | AAB81135.1 | |
| U95970.1 | 7 | 975 | AAD00821.1 |
More Information
Set Membership: |
|

Genomic Map