Assay Details
Target Gene Details
Entrez Gene ID: | 51406 |
Gene Name: | nucleolar protein 7 |
Gene Aliases: |
C6orf90, PQBP3, RARG-1, dJ223E5.2 |
Location: |
Chr.6:13615327-13632470 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOL7 | NM_001317724.1 | NP_001304653.1 | ||
| BU071935.1 |
Target Gene Details
Entrez Gene ID: | 10048 |
Gene Name: | RAN binding protein 9 |
Gene Aliases: |
BPM-L, BPM90, RANBPM, RanBP7 |
Location: |
Chr.6:13621498-13711582 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RANBP9 | NM_005493.2 | 14 | 2822 | NP_005484.2 |
| XM_006714945.2 | 14 | 2302 | XP_006715008.1 | |
| XM_011514205.2 | 12 | 2570 | XP_011512507.1 | |
| XM_017010149.1 | 14 | 2210 | XP_016865638.1 | |
| AF306510.1 | 14 | 2360 | AAK15469.1 | |
| BC052781.1 | 14 | 2983 | AAH52781.1 | |
| BC063849.1 | 12 | 2078 | AAH63849.1 |
More Information
Set Membership: |
|

Genomic Map