Assay Details
Target Gene Details
Entrez Gene ID: | 3552 |
Gene Name: | interleukin 1 alpha |
Gene Aliases: |
IL-1A, IL1, IL1-ALPHA, IL1F1 |
Location: |
Chr.2:112773915-112785398 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1A | NM_000575.4 | 1 | 740 | NP_000566.3 |
More Information
Set Membership: |
|

Genomic Map