Assay Details
Target Gene Details
Entrez Gene ID: | 285761 |
Gene Name: | discoidin, CUB and LCCL domain containing 1 |
Gene Aliases: |
dJ94G16.1 |
Location: |
Chr.6:117480346-117569858 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCBLD1 | NM_173674.2 | 15 | 1915 | NP_775945.1 |
| BC035671.1 | 15 | 1862 | AAH35671.1 | |
| CR749448.1 | 11 | 3467 | CAH18285.1 |
Target Gene Details
Entrez Gene ID: | 57120 |
Gene Name: | golgi associated PDZ and coiled-coil motif containing |
Gene Aliases: |
CAL, FIG, GOPC1, PIST, dJ94G16.2 |
Location: |
Chr.6:117560269-117602542 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOPC | NM_001017408.2 | NP_001017408.1 | ||
| NM_020399.3 | NP_065132.1 | |||
| AB209385.1 | 6 | 2231 | BAD92622.1 | |
| AF287894.1 | AAG00572.1 | |||
| AF450008.1 | AAL47160.1 | |||
| AK314486.1 | ||||
| AY033606.1 | AAK57733.1 | |||
| BC009553.2 | AAH09553.1 | |||
| KF420122.1 | ||||
| KF420123.1 |
More Information
Set Membership: |
|

Genomic Map