Assay Details
Target Gene Details
Entrez Gene ID: | 3316 |
Gene Name: | heat shock protein family B (small) member 2 |
Gene Aliases: |
HSP27, Hs.78846, LOH11CR1K, MKBP |
Location: |
Chr.11:111912736-111914093 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPB2 | NM_001541.3 | 2 | 662 | NP_001532.1 |
| AB096250.1 | 2 | 1156 | ||
| AI131288.1 | 1 | 197 | ||
| BC109393.1 | 2 | 597 | AAI09394.1 | |
| CR457408.1 | CAG33689.1 | |||
| D89617.1 | 2 | 637 | BAA24004.1 | |
| S67070.1 | 1 | 307 | AAB28816.1 |
Target Gene Details
Entrez Gene ID: | 100528019 |
Gene Name: | HSPB2-C11orf52 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:111912736-111926871 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPB2-C11orf52 | NR_037651.1 | |||
| AK293041.1 | 2 | 662 | ||
| BC110872.1 | AAI10873.1 |
More Information
Set Membership: |
|

Genomic Map