Assay Details
Target Gene Details
Entrez Gene ID: | 7158 |
Gene Name: | tumor protein p53 binding protein 1 |
Gene Aliases: |
53BP1, TDRD30, TP53, p202, p53BP1 |
Location: |
Chr.15:43403064-43510509 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP53BP1 | NM_001141979.1 | 22 | 4892 | NP_001135451.1 |
| NM_001141980.1 | 22 | 4892 | NP_001135452.1 | |
| NM_005657.2 | 22 | 4912 | NP_005648.1 | |
| XM_011521985.1 | 21 | 4741 | XP_011520287.1 | |
| XM_011521986.2 | 22 | 4891 | XP_011520288.1 | |
| XM_017022533.1 | 23 | 4996 | XP_016878022.1 | |
| AB210025.1 | 22 | 4863 | ||
| AF078776.1 | 22 | 4937 | AAC62018.1 | |
| AK123418.1 | 1 | 898 | ||
| AK295764.1 | 22 | 4892 | ||
| BC112161.1 | 22 | 4872 | AAI12162.1 | |
| BX537418.1 | 22 | 4891 | CAD97660.1 | |
| U09477.1 | 9 | 1931 | AAA21596.1 |
More Information
Set Membership: |
|

Genomic Map