Assay Details
Target Gene Details
Entrez Gene ID: | 100529067 |
Gene Name: | SERF2-C15orf63 readthrough |
Gene Aliases: |
- |
Location: |
Chr.15:43791976-43802589 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERF2-C15ORF63 | NR_037673.1 | |||
| AF370428.1 | 1 | 2903 | AAQ15264.1 | |
| AK000438.1 | BAA91165.1 | |||
| AK094716.1 | 1 | 737 | ||
| AK095876.1 | 3 | 314 |
Target Gene Details
Entrez Gene ID: | 619189 |
Gene Name: | serine incorporator 4 |
Gene Aliases: |
- |
Location: |
Chr.15:43794174-43800097 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERINC4 | NM_001258031.1 | 3 | 485 | NP_001244960.1 |
| NM_001258032.1 | 3 | 526 | NP_001244961.1 | |
| AK097682.1 | 1 | 91 | ||
| BC136669.1 | 1 | 322 | ||
| BX647438.1 | 3 | 526 | ||
| DQ103711.1 | 3 | 486 | AAZ80298.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1036318 | Chr.15:43761419 - 43868575 on Build GRCh38 | Gain |
|
| esv2760028 | Chr.15:43530960 - 43856443 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map