Assay Details
Target Gene Details
Entrez Gene ID: | 22885 |
Gene Name: | actin binding LIM protein family member 3 |
Gene Aliases: |
HMFN1661 |
Location: |
Chr.5:149141483-149260439 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABLIM3 | NM_001301015.1 | 1 | 545 | NP_001287944.1 |
| NM_001301018.1 | NP_001287947.1 | |||
| NM_001301027.1 | 1 | 545 | NP_001287956.1 | |
| NM_001301028.1 | 1 | 545 | NP_001287957.1 | |
| NM_014945.3 | NP_055760.1 | |||
| XM_005268392.1 | XP_005268449.1 | |||
| XM_005268393.1 | XP_005268450.1 | |||
| XM_005268394.1 | XP_005268451.1 | |||
| XM_005268395.1 | XP_005268452.1 | |||
| XM_005268396.1 | XP_005268453.1 | |||
| XM_011537609.2 | XP_011535911.1 | |||
| XM_017009238.1 | 1 | 266 | XP_016864727.1 | |
| AB020650.1 | ||||
| AK225769.1 | 1 | 414 | ||
| AK289736.1 | ||||
| BC001665.2 | 1 | 207 | AAH01665.1 | |
| BP285049.1 | ||||
| DQ413174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830517 | Chr.5:148953659 - 149171578 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map