Assay Details
Target Gene Details
Entrez Gene ID: | 3750 |
Gene Name: | potassium voltage-gated channel subfamily D member 1 |
Gene Aliases: |
KV4.1 |
Location: |
Chr.X:48960983-48972099 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCND1 | NM_004979.5 | 6 | 4253 | NP_004970.3 |
| XM_011543910.2 | 7 | 3161 | XP_011542212.1 | |
| XM_017029508.1 | 7 | 2992 | XP_016884997.1 | |
| XM_017029509.1 | 7 | 2989 | XP_016884998.1 | |
| AB021865.1 | BAA96454.1 | |||
| AJ005898.1 | 6 | 2680 | CAA06755.1 | |
| AK094431.1 | 5 | 2839 | ||
| BC045659.1 | 6 | 4251 | AAH45659.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map