Assay Details
Target Gene Details
Entrez Gene ID: | 57541 |
Gene Name: | zinc finger protein 398 |
Gene Aliases: |
P51, P71, ZER6 |
Location: |
Chr.7:149126416-149183042 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF398 | NM_020781.3 | 7 | 3090 | NP_065832.1 |
| NM_170686.2 | 6 | 3027 | NP_733787.1 | |
| XM_011516439.2 | 7 | 2908 | XP_011514741.1 | |
| XM_011516440.2 | 6 | 2880 | XP_011514742.1 | |
| XM_011516441.2 | 6 | 2863 | XP_011514743.1 | |
| AB037760.1 | 2 | 2053 | BAA92577.1 | |
| AK026380.1 | ||||
| AY049743.1 | 7 | 3090 | AAK92788.1 | |
| AY049744.1 | 6 | 2769 | AAK92789.1 |
More Information
Set Membership: |
|

Genomic Map