Assay Details
Target Gene Details
Entrez Gene ID: | 4947 |
Gene Name: | ornithine decarboxylase antizyme 2 |
Gene Aliases: |
AZ2 |
Location: |
Chr.15:64687574-64703281 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OAZ2 | NM_001301302.1 | 5 | 795 | NP_001288231.1 |
| NM_002537.3 | 5 | 798 | NP_002528.1 | |
| AF057297.1 | 5 | 716 | AAD03265.1 | |
| AF242520.1 | 5 | 693 | ||
| AF242521.1 | 5 | 692 | AAF99601.1 | |
| BI597670.1 | 5 | 733 | ||
| BX648172.1 | 4 | 5955 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833038 | Chr.15:64522753 - 64724042 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map