Assay Details
Target Gene Details
Entrez Gene ID: | 6426 |
Gene Name: | serine and arginine rich splicing factor 1 |
Gene Aliases: |
ASF, SF2, SF2p33, SFRS1, SRp30a |
Location: |
Chr.17:58000919-58007346 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF1 | NM_001078166.1 | 3 | 1638 | NP_001071634.1 |
| NM_006924.4 | 4 | 1438 | NP_008855.1 | |
| NR_034041.1 | ||||
| AB062124.1 | 4 | 1319 | BAB93456.1 | |
| AB209558.1 | 3 | 1525 | BAD92795.1 | |
| AK225711.1 | ||||
| AK289933.1 | ||||
| AK298527.1 | 3 | 1020 | ||
| BC010264.1 | 4 | 1264 | AAH10264.1 | |
| M69040.1 | 4 | 1353 | AAA03476.1 | |
| M72709.1 | AAA35564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952347 | Chr.17:57999240 - 58005339 on Build GRCh38 | Duplication |
|
| nsv833497 | Chr.17:57929210 - 58108015 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map