Assay Details
Target Gene Details
Entrez Gene ID: | 5048 |
Gene Name: | platelet activating factor acetylhydrolase 1b regulatory subunit 1 |
Gene Aliases: |
LIS1, LIS2, MDCR, MDS, NudF, PAFAH |
Location: |
Chr.17:2593210-2685617 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAFAH1B1 | NM_000430.3 | 1 | 169 | NP_000421.1 |
| XM_011523901.1 | 1 | 164 | XP_011522203.1 | |
| XM_017024701.1 | XP_016880190.1 | |||
| L13386.1 | AAA02881.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1070794 | Chr.17:2593605 - 2594206 on Build GRCh38 | Deletion |
|
| nsv833339 | Chr.17:2578764 - 2769775 on Build GRCh38 | Loss |
|
| nsv1055844 | Chr.17:2586358 - 2635403 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map