Assay Details
Target Gene Details
Entrez Gene ID: | 6295 |
Gene Name: | S-antigen visual arrestin |
Gene Aliases: |
RP47, S-AG |
Location: |
Chr.2:233307663-233347066 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAG | NM_000541.4 | NP_000532.2 | ||
| AA317634.1 | ||||
| BQ639460.1 | ||||
| BX491422.1 | ||||
| BX641043.1 | CAE46025.1 | |||
| BX647821.1 | ||||
| BX647827.1 | ||||
| DQ980618.1 | ||||
| DQ980619.1 | ||||
| DQ980620.1 | ||||
| X12453.1 | CAA30983.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584699 | Chr.2:233273594 - 233346901 on Build GRCh38 | Gain |
|
| nsv584700 | Chr.2:233290755 - 233455780 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map