Assay Details
Target Gene Details
Entrez Gene ID: | 1582 |
Gene Name: | cytochrome P450 family 8 subfamily B member 1 |
Gene Aliases: |
CP8B, CYP12 |
Location: |
Chr.3:42872192-42876141 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP8B1 | NM_004391.2 | 1 | 2186 | NP_004382.2 |
| AF090318.1 | 1 | 1745 | AAC63037.1 | |
| AK310784.1 |
More Information
Set Membership: |
|

Genomic Map