Assay Details
Target Gene Details
Entrez Gene ID: | 10658 |
Gene Name: | CUGBP, Elav-like family member 1 |
Gene Aliases: |
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50 |
Location: |
Chr.11:47465937-47565569 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832141 | Chr.11:47311629 - 47499441 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map