Assay Details
Target Gene Details
Entrez Gene ID: | 1262 |
Gene Name: | cyclic nucleotide gated channel alpha 4 |
Gene Aliases: |
CNCA2, CNG-4, CNG4, CNG5, CNGB2, OCNC2, OCNCBETA, OCNCb |
Location: |
Chr.11:6234774-6245924 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNGA4 | NM_001037329.3 | NP_001032406.1 | ||
| XM_017017217.1 | XP_016872706.1 | |||
| XM_017017218.1 | XP_016872707.1 | |||
| XM_017017219.1 | XP_016872708.1 | |||
| AK122736.1 | ||||
| AK302551.1 | ||||
| BC040277.1 | AAH40277.1 | |||
| DB238337.1 |
More Information
Set Membership: |
|

Genomic Map