Assay Details
Target Gene Details
Entrez Gene ID: | 4047 |
Gene Name: | lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase) |
Gene Aliases: |
CTRCT44, OSC |
Location: |
Chr.21:46188446-46228824 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LSS | NM_001001438.2 | NP_001001438.1 | ||
| NM_001145436.1 | 22 | 2596 | NP_001138908.1 | |
| NM_001145437.1 | 21 | 2761 | NP_001138909.1 | |
| NM_002340.5 | 22 | 2629 | NP_002331.3 | |
| XM_011529564.2 | 22 | 2655 | XP_011527866.1 | |
| XM_011529565.1 | XP_011527867.1 | |||
| XM_011529566.2 | 22 | 2576 | XP_011527868.1 | |
| XM_011529567.2 | 21 | 2559 | XP_011527869.1 | |
| XM_017028346.1 | XP_016883835.1 | |||
| XM_017028347.1 | XP_016883836.1 | |||
| XM_017028348.1 | XP_016883837.1 | |||
| AK092334.1 | 3 | 1979 | BAC03865.1 | |
| AK096769.1 | BAC04859.1 | |||
| AK098352.1 | 2 | 117 | BAC05289.1 | |
| AK226141.1 | 21 | 2674 | ||
| AK296313.1 | ||||
| BC035638.1 | 22 | 2592 | AAH35638.1 | |
| BU728696.1 | ||||
| D63807.1 | BAA09875.1 | |||
| S81221.1 | AAB36220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2338420 | Chr.21:46189668 - 46191124 on Build GRCh38 | Deletion |
|
| nsv519904 | Chr.21:46163592 - 46461396 on Build GRCh38 | Loss |
|
| nsv1061087 | Chr.21:46056174 - 46223305 on Build GRCh38 | Loss |
|
| nsv509804 | Chr.21:45879599 - 46235140 on Build GRCh38 | Insertion |
|
| nsv1062031 | Chr.21:46188666 - 46223305 on Build GRCh38 | Gain |
|
| esv29868 | Chr.21:46189865 - 46190920 on Build GRCh38 | Loss |
|
| nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain |
|
| esv2587417 | Chr.21:46188898 - 46191269 on Build GRCh38 | Deletion |
|
| nsv1060978 | Chr.21:46068159 - 46413726 on Build GRCh38 | Gain |
|
| esv3647164 | Chr.21:46189814 - 46191090 on Build GRCh38 | Loss |
|
| esv2668812 | Chr.21:46189828 - 46190962 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map