Assay Details
Target Gene Details
Entrez Gene ID: | 55974 |
Gene Name: | solute carrier family 50 member 1 |
Gene Aliases: |
HsSWEET1, RAG1AP1, SCP, SWEET1, slv |
Location: |
Chr.1:155135253-155138858 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
| nsv547961 | Chr.1:155109822 - 155225189 on Build GRCh38 | Loss |
|
| nsv998756 | Chr.1:155107614 - 155164069 on Build GRCh38 | Gain |
|
| esv3587615 | Chr.1:155047853 - 155175013 on Build GRCh38 | Gain |
|
| dgv3n68 | Chr.1:155122502 - 155343618 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map