Assay Details
Target Gene Details
Entrez Gene ID: | 101929057 |
Gene Name: | uncharacterized LOC101929057 |
Gene Aliases: |
- |
Location: |
Chr.6:89638532-89689481 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929057 | NR_111915.1 | |||
| CR936797.1 |
Target Gene Details
Entrez Gene ID: | 23195 |
Gene Name: | midasin AAA ATPase 1 |
Gene Aliases: |
Rea1 |
Location: |
Chr.6:89642775-89828021 on Build GRCh38 |
Assay Gene Location: | Within Exon 93 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MDN1 | NM_014611.2 | 91 | 15414 | NP_055426.1 |
| XM_005248700.3 | 52 | 9346 | XP_005248757.1 | |
| XM_006715405.3 | 55 | 10892 | XP_006715468.1 | |
| XM_011535635.2 | 91 | 15219 | XP_011533937.1 | |
| XM_011535636.2 | 91 | 15384 | XP_011533938.1 | |
| AB002299.2 | 45 | 8070 | BAA20761.2 | |
| AF503925.1 | 91 | 15343 | AAM77722.1 | |
| CR627399.1 | 37 | 6869 | CAH10492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604124 | Chr.6:89552180 - 89685297 on Build GRCh38 | Loss |
|
| esv3890901 | Chr.6:89204468 - 90033564 on Build GRCh38 | Gain |
|
| dgv6089n100 | Chr.6:89446386 - 89678417 on Build GRCh38 | Gain |
|
| esv3609903 | Chr.6:89446569 - 89685111 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map