Assay Details
Target Gene Details
Entrez Gene ID: | 5603 |
Gene Name: | mitogen-activated protein kinase 13 |
Gene Aliases: |
MAPK 13, MAPK-13, PRKM13, SAPK4, p38delta |
Location: |
Chr.6:36130484-36144524 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK13 | NM_002754.4 | NP_002745.1 | ||
| NR_072996.1 | ||||
| AB209586.1 | BAD92823.1 | |||
| AB451221.1 | ||||
| AB451344.1 | ||||
| AF004709.1 | AAC51374.1 | |||
| AF015256.1 | AAC51758.1 | |||
| AF092535.1 | AAD23377.1 | |||
| AF100546.1 | AAF36772.1 | |||
| AK222922.1 | BAD96642.1 | |||
| AK308018.1 | ||||
| BC000433.2 | AAH00433.1 | |||
| BC001641.2 | AAH01641.1 | |||
| BC004428.1 | AAH04428.1 | |||
| BC085196.1 | AAH85196.1 | |||
| BM921729.1 | ||||
| BT007221.1 | AAP35885.1 | |||
| CR536490.1 | CAG38729.1 | |||
| CR541687.1 | CAG46488.1 | |||
| KU178281.1 | ||||
| KU178282.1 | ||||
| U93232.1 | AAB87639.1 | |||
| Y10488.1 | CAA71512.1 |
More Information
Set Membership: |
|

Genomic Map