Assay Details
Target Gene Details
Entrez Gene ID: | 78996 |
Gene Name: | chromosome 7 open reading frame 49 |
Gene Aliases: |
MRI |
Location: |
Chr.7:135092303-135170826 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C7orf49 | NM_001305630.1 | NP_001292559.1 | ||
| XM_017012589.1 | 4 | 2322 | XP_016868078.1 | |
| XM_017012590.1 | 4 | 2248 | XP_016868079.1 | |
| XM_017012591.1 | 4 | 2102 | XP_016868080.1 | |
| XM_017012592.1 | 3 | 2283 | XP_016868081.1 | |
| XM_017012593.1 | 4 | 2152 | XP_016868082.1 | |
| XM_017012594.1 | 4 | 2359 | XP_016868083.1 | |
| XM_017012595.1 | XP_016868084.1 |
Target Gene Details
Entrez Gene ID: | 55281 |
Gene Name: | transmembrane protein 140 |
Gene Aliases: |
- |
Location: |
Chr.7:135148014-135166215 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM140 | NM_018295.4 | 2 | 1039 | NP_060765.4 |
| AK001862.1 | 2 | 981 | BAA91948.1 | |
| AK225055.1 | ||||
| AK301268.1 | 3 | 855 | ||
| BC020942.1 | 2 | 966 | AAH20942.1 | |
| BM512090.1 | 1 | 296 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527014 | Chr.7:135149118 - 135206907 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map