Assay Details
Target Gene Details
Entrez Gene ID: | 57030 |
Gene Name: | solute carrier family 17 member 7 |
Gene Aliases: |
BNPI, VGLUT1 |
Location: |
Chr.19:49429398-49441551 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC17A7 | NM_020309.3 | NP_064705.1 | ||
| AB032436.1 | BAA92875.1 | |||
| AK091061.1 | ||||
| AK289622.1 | ||||
| AK294226.1 | ||||
| AK294405.1 | ||||
| BC059379.1 | AAH59379.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| dgv52n68 | Chr.19:49286685 - 49456131 on Build GRCh38 | Loss |
|
| nsv953599 | Chr.19:49386444 - 49506743 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map