Assay Details
Target Gene Details
Entrez Gene ID: | 11100 |
Gene Name: | heterogeneous nuclear ribonucleoprotein U like 1 |
Gene Aliases: |
E1B-AP5, E1BAP5, HNRPUL1 |
Location: |
Chr.19:41262476-41307783 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPUL1 | NM_001301016.2 | 2 | 373 | NP_001287945.1 |
| NM_001321208.1 | 2 | 413 | NP_001308137.1 | |
| NM_001321211.1 | 2 | 197 | NP_001308140.1 | |
| NM_007040.5 | 2 | 704 | NP_008971.2 | |
| NM_144732.4 | 2 | 186 | NP_653333.1 | |
| XM_005258459.3 | 2 | 507 | XP_005258516.1 | |
| XM_005258461.4 | 2 | 337 | XP_005258518.1 | |
| XM_005258463.3 | 2 | 328 | XP_005258520.1 | |
| XM_005258464.3 | 2 | 143 | XP_005258521.1 | |
| XM_005258465.4 | 2 | 354 | XP_005258522.1 | |
| XM_011526392.2 | 2 | 507 | XP_011524694.1 | |
| XM_011526393.2 | 2 | 429 | XP_011524695.1 | |
| XM_011526394.2 | 2 | 184 | XP_011524696.1 | |
| XM_011526395.2 | 2 | 393 | XP_011524697.1 | |
| AJ007509.1 | 2 | 547 | CAA07548.1 | |
| AK022863.1 | 2 | 506 | ||
| AK023006.1 | 2 | 143 | ||
| AK127057.1 | 3 | 377 | BAC86806.1 | |
| AK225850.1 | 2 | 166 | ||
| AK290729.1 | 2 | 168 | ||
| AK291315.1 | 2 | 453 | ||
| AK291762.1 | 2 | 506 | ||
| AK297127.1 | 2 | 301 | ||
| AK308078.1 | 2 | 434 | ||
| AK308156.1 | 3 | 187 | ||
| BC002564.2 | 2 | 444 | AAH02564.1 | |
| BC004242.1 | AAH04242.1 | |||
| BC009988.2 | 2 | 350 | AAH09988.2 | |
| BC014232.2 | 2 | 325 | AAH14232.1 | |
| BC027713.2 | 2 | 427 | AAH27713.1 | |
| DC347248.1 | 2 | 187 | ||
| DC366321.1 | 2 | 176 | ||
| DC403921.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26941 | Chr.19:41264323 - 41309377 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map