Assay Details
Target Gene Details
Entrez Gene ID: | 6748 |
Gene Name: | signal sequence receptor subunit 4 |
Gene Aliases: |
CDG1Y, TRAPD |
Location: |
Chr.X:153794175-153798512 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSR4 | NM_001204526.1 | NP_001191455.1 | ||
| NM_001204527.1 | NP_001191456.1 | |||
| NM_006280.2 | NP_006271.1 | |||
| NR_037927.1 | ||||
| XM_011531186.1 | XP_011529488.1 | |||
| XM_011531187.1 | XP_011529489.1 | |||
| XM_017029756.1 | XP_016885245.1 | |||
| XM_017029757.1 | XP_016885246.1 | |||
| AK057117.1 | ||||
| AK094395.1 | 3 | 1089 | ||
| AK290493.1 | ||||
| BC003371.1 | AAH03371.1 | |||
| BE791319.1 | ||||
| BG165852.1 | ||||
| BP202705.1 | ||||
| BT007192.1 | AAP35856.1 | |||
| CD672615.1 | ||||
| DB238237.1 | ||||
| X90583.1 | CAA62211.1 | |||
| Z69043.1 | CAA93157.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|

Genomic Map