Assay Details
Target Gene Details
Entrez Gene ID: | 4485 |
Gene Name: | macrophage stimulating 1 |
Gene Aliases: |
D3F15S2, DNF15S2, HGFL, MSP, NF15S2 |
Location: |
Chr.3:49683947-49689528 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MST1 | NM_020998.3 | NP_066278.3 | ||
| XM_006713166.1 | XP_006713229.1 | |||
| XM_011533732.1 | XP_011532034.1 | |||
| XM_017006460.1 | XP_016861949.1 | |||
| XM_017006461.1 | XP_016861950.1 | |||
| XM_017006462.1 | XP_016861951.1 | |||
| XM_017006463.1 | XP_016861952.1 | |||
| XM_017006464.1 | XP_016861953.1 | |||
| AK222893.1 | BAD96613.1 | |||
| BC048330.1 | AAH48330.1 | |||
| BM672747.1 | ||||
| L11924.1 | AAA59872.1 | |||
| M74178.1 | AAA50165.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471425 | Chr.3:49683948 - 49688763 on Build GRCh38 | Gain |
|
| nsv1148681 | Chr.3:49683166 - 49692167 on Build GRCh38 | Duplication |
|
| esv28815 | Chr.3:49683318 - 49698418 on Build GRCh38 | Gain+Loss |
|
| nsv1141429 | Chr.3:49676267 - 49693067 on Build GRCh38 | Duplication |
|
| nsv822077 | Chr.3:49683967 - 49697093 on Build GRCh38 | Gain |
|
| nsv508217 | Chr.3:49578013 - 49732463 on Build GRCh38 | Deletion |
|
| nsv820990 | Chr.3:49683218 - 49698478 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map