Assay Details
Target Gene Details
Entrez Gene ID: | 816 |
Gene Name: | calcium/calmodulin dependent protein kinase II beta |
Gene Aliases: |
CAM2, CAMK2, CAMKB |
Location: |
Chr.7:44217150-44325994 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 23 - Intron 24 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv606752 | Chr.7:44220272 - 44227535 on Build GRCh38 | Gain |
|
| nsv606751 | Chr.7:44213422 - 44232814 on Build GRCh38 | Gain |
|
| nsv509195 | Chr.7:44147118 - 44261782 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map