Assay Details
Target Gene Details
Entrez Gene ID: | 10325 |
Gene Name: | Ras related GTP binding B |
Gene Aliases: |
RAGB, bA465E19.1 |
Location: |
Chr.X:55717677-55760209 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRAGB | NM_006064.4 | NP_006055.3 | ||
| NM_016656.3 | NP_057740.2 | |||
| XM_005261973.4 | XP_005262030.1 | |||
| XM_017029217.1 | XP_016884706.1 | |||
| XM_017029218.1 | XP_016884707.1 | |||
| AK289407.1 | ||||
| AL831926.1 | CAD38586.1 | |||
| BC034726.1 | AAH34726.1 | |||
| X90530.1 | CAA62132.1 |
More Information
Set Membership: |
|

Genomic Map