Assay Details
Target Gene Details
Entrez Gene ID: | 101927051 |
Gene Name: | uncharacterized LOC101927051 |
Gene Aliases: |
- |
Location: |
Chr.22:37641832-37658377 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927051 | NR_109952.1 | |||
| AK097791.1 |
Target Gene Details
Entrez Gene ID: | 23616 |
Gene Name: | SH3 domain binding protein 1 |
Gene Aliases: |
ARHGAP43 |
Location: |
Chr.22:37639677-37656043 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3BP1 | NM_018957.3 | NP_061830.3 | ||
| AK124370.1 | BAC85842.1 | |||
| AK126556.1 | BAC86592.1 | |||
| AK301163.1 | ||||
| AY364241.1 | AAQ76800.1 | |||
| BC008282.1 | AAH08282.1 | |||
| CR456576.1 | CAG30462.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829203 | Chr.22:36998126 - 37773322 on Build GRCh38 | Loss |
|
| esv2758838 | Chr.22:37607870 - 37869871 on Build GRCh38 | Loss |
|
| nsv589006 | Chr.22:37642053 - 37658606 on Build GRCh38 | Loss |
|
| nsv471197 | Chr.22:37600773 - 37762734 on Build GRCh38 | Loss |
|
| nsv834191 | Chr.22:37533773 - 37717344 on Build GRCh38 | Loss |
|
| nsv1057617 | Chr.22:37462633 - 37713698 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74516928] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map