Assay Details
Target Gene Details
Entrez Gene ID: | 83873 |
Gene Name: | G protein-coupled receptor 61 |
Gene Aliases: |
BALGR, GPCR3 |
Location: |
Chr.1:109537539-109547260 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR61 | NM_031936.4 | 3 | 2517 | NP_114142.3 |
| XM_017002438.1 | 2 | 5521 | XP_016857927.1 | |
| AF258342.1 | 4 | 2153 | AAK97826.1 |
More Information
Set Membership: |
|

Genomic Map